3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride 97%
Dodavatel: Apollo Scientific
Synonyms:
3-(Trifluoromethyl)-3-pyrrolidinol hydrochloride, 3-(Trifluoromethyl)pyrrolidin-3-ol hydrochloride
PC530026-1G
APOSPC530026-1GEA
5890
CZK
APOSPC530026-1G
3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride 97%
3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride
Vzorec:
C₅H₉ClF₃NO MW: 191,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19982573 CAS číslo: 1334147-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...