Tisk…
1-(4-Fluorophenyl)cyclohexanecarboxylic acid 97%
Dodavatel: Apollo Scientific
Synonyms: 1-(4-Fluorophenyl)cyclohexane-1-carboxylic acid
PC530023-1G PC530023-250MG
APOSPC530023-1GEA 6190 CZK
APOSPC530023-1G APOSPC530023-250MG
1-(4-Fluorophenyl)cyclohexanecarboxylic acid 97%
1-(4-Fluorophenyl)cyclohexanecarboxylic acid
Vzorec: C₁₃H₁₅FO₂
MW: 222,2554 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800627
CAS číslo: 214263-00-0