1-(3-Fluorophenyl)cyclopentanecarboxylic acid 97%
Dodavatel: Apollo Scientific
PC530022-1G
PC530022-5G
APOSPC530022-1GEA
857
CZK
APOSPC530022-1G
APOSPC530022-5G
1-(3-Fluorophenyl)cyclopentanecarboxylic acid 97%
1-(3-Fluorophenyl)cyclopentanecarboxylic acid
Vzorec:
C₁₂H₁₃FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00800624 CAS číslo: 214262-97-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...