Tisk…

H-D-Phe(3,5-DiF)-OH 97%

Dodavatel: Apollo Scientific

PC530000-1G PC530000-5G
APOSPC530000-1GEA 3150 CZK
APOSPC530000-1G APOSPC530000-5G
H-D-Phe(3,5-DiF)-OH 97%
H-D-Phe(3,5-DiF)-OH
Vzorec: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD01631988
CAS číslo: 266360-63-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR