Tisk…

Nitrosium hexafluorophosphate 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Nitrosonium hexafluorophosphate, Nitrosyl hexafluorophosphate

PC5280-25G PC5280-5G
APOSPC5280-25GEA 19740 CZK
APOSPC5280-25G APOSPC5280-5G
Nitrosium hexafluorophosphate 96%
Nitrosium hexafluorophosphate
Vzorec: F₆NOP
MW: 174,97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040324
CAS číslo: 16921-91-8
UN: 3260
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR