Tisk…

2-Fluoro-3-nitrobenzotrifluoride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC5265-5G PC5265-1G PC5265-25G
APOSPC5265-5GEA 2860 CZK
APOSPC5265-5G APOSPC5265-1G APOSPC5265-25G
2-Fluoro-3-nitrobenzotrifluoride 98%
2-Fluoro-3-nitrobenzotrifluoride
Vzorec: C₇H₃F₄NO₂
MW: 209,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08532487
CAS číslo: 61324-97-8
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR