Nitronium hexafluoroantimonate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC5258-100G
PC5258-25G
APOSPC5258-100GEA
21430
CZK
APOSPC5258-100G
APOSPC5258-25G
Nitronium hexafluoroantimonate 98%
Nitronium hexafluoroantimonate
Vzorec:
F₆NO₂Sb Storage Temperature: Refrigerator |
MDL Number:
MFCD00042551 CAS číslo: 17856-92-7 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...