Tisk…

2,6-Dichloro-4-(trifluoromethoxy)benzoic acid 97%

Dodavatel: Apollo Scientific

PC52461-1G PC52461-5G
APOSPC52461-1GEA 2150 CZK
APOSPC52461-1G APOSPC52461-5G
2,6-Dichloro-4-(trifluoromethoxy)benzoic acid 97%
2,6-Dichloro-4-(trifluoromethoxy)benzoic acid
Vzorec: C₈H₃Cl₂F₃O₃
MW: 275,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660297
CAS číslo: 886502-90-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR