Tisk…

5-Fluoropicolinic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Fluoro-2-pyridinecarboxylic acid, 5-Fluoro-2-picolinic acid, 5-Fluoropyridine-2-carboxylic acid

PC5246-25G PC5246-5G PC5246-100G
APOSPC5246-25GEA 2110 CZK
APOSPC5246-25G APOSPC5246-5G APOSPC5246-100G
5-Fluoropicolinic acid 98%
5-Fluoropicolinic acid
Vzorec: C₆H₄FNO₂
MW: 141,1 g/mol
Bod tání: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD04114196
CAS číslo: 107504-08-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR