Methyl-3-trifluoromethylbenzoylacetate 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
PC5234-1G
PC5234-5G
APOSPC5234-1GEA
1820
CZK
APOSPC5234-1G
APOSPC5234-5G
Methyl-3-trifluoromethylbenzoylacetate 95+%
Methyl-3-trifluoromethylbenzoylacetate
Vzorec:
C₁₁H₉F₃O₃ MW: 246,185 g/mol Bod varu: 94…96 °C/0,2mmHg Bod vzplanutí: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216522 CAS číslo: 93618-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...