2-(2-Fluorophenyl)-4-methylthiazole-5-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC52223-1G
PC52223-250MG
APOSPC52223-1GEA
10980
CZK
APOSPC52223-1G
APOSPC52223-250MG
2-(2-Fluorophenyl)-4-methylthiazole-5-carboxylic acid
2-(2-Fluorophenyl)-4-methylthiazole-5-carboxylic acid
Vzorec:
C₁₁H₈FNO₂S MW: 237,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06857924 CAS číslo: 879070-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...