Bis(pentafluorophenyl)carbonate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Bis(pentafluorophenyl) Carbonate
PC5218-25G
PC5218-100G
PC5218-5G
APOSPC5218-25GEA
1950
CZK
APOSPC5218-25G
APOSPC5218-100G
APOSPC5218-5G
Bis(pentafluorophenyl)carbonate 98%
Bis(pentafluorophenyl)carbonate
Vzorec:
C₁₃F₁₀O₃ MW: 394.13 Bod tání: 47…51 °C Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00368353 CAS číslo: 59483-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...