5-Bromo-2,4-difluorobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC52118-1G
PC52118-25G
PC52118-5G
APOSPC52118-1GEA
622
CZK
APOSPC52118-1G
APOSPC52118-25G
APOSPC52118-5G
5-Bromo-2,4-difluorobenzaldehyde
5-Bromo-2,4-difluorobenzaldehyde
Vzorec:
C₇H₃BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD09842439 CAS číslo: 473416-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...