Tisk…

5-Bromo-2,4-bis(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific

PC520908-1G PC520908-5G
APOSPC520908-1GEA 8040 CZK
APOSPC520908-1G APOSPC520908-5G
5-Bromo-2,4-bis(trifluoromethyl)pyridine 95%
5-Bromo-2,4-bis(trifluoromethyl)pyridine
Vzorec: C₇H₂BrF₆N
Storage Temperature: Ambient
MDL Number: MFCD11100546
CAS číslo: 1099598-05-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR