Tisk…

2-Chloro-5-fluoro-3-(trifluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific

PC520903-1G PC520903-5G
APOSPC520903-1GEA 4860 CZK
APOSPC520903-1G APOSPC520903-5G
2-Chloro-5-fluoro-3-(trifluoromethyl)pyridine 97%
2-Chloro-5-fluoro-3-(trifluoromethyl)pyridine
Vzorec: C₆H₂ClF₄N
Storage Temperature: Ambient
MDL Number: MFCD11100544
CAS číslo: 72587-20-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR