Tisk…

1-Fluoro-2,3-bis(trifluoromethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520816-1G PC520816-500MG PC520816-5G
APOSPC520816-1GEA 6480 CZK
APOSPC520816-1G APOSPC520816-500MG APOSPC520816-5G
1-Fluoro-2,3-bis(trifluoromethyl)benzene 95%
1-Fluoro-2,3-bis(trifluoromethyl)benzene
Vzorec: C₈H₃F₇
MW: 232,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226555
CAS číslo: 1099597-20-2
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR