Tisk…

2,3-Difluoro-1,4-bis(trifluoromethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520814-1G PC520814-500MG PC520814-5G
APOSPC520814-1GEA 9920 CZK
APOSPC520814-1G APOSPC520814-500MG APOSPC520814-5G
2,3-Difluoro-1,4-bis(trifluoromethyl)benzene 95%
2,3-Difluoro-1,4-bis(trifluoromethyl)benzene
Vzorec: C₈H₂F₈
MW: 250,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226683
CAS číslo: 1099597-67-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR