Tisk…

1-Fluoro-4-nitro-2-(2,2,2-trifluoroethyl)-benzene 95%

Dodavatel: Apollo Scientific

PC520801-10G PC520801-1G PC520801-5G
APOSPC520801-10GEA 39350 CZK
APOSPC520801-10G APOSPC520801-1G APOSPC520801-5G
1-Fluoro-4-nitro-2-(2,2,2-trifluoroethyl)-benzene 95%
1-Fluoro-4-nitro-2-(2,2,2-trifluoroethyl)-benzene
Vzorec: C₈H₅F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD18384827
CAS číslo: 1262413-56-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR