Tisk…

2,3-Dimethylbenzotrifluoride 97%

Dodavatel: Apollo Scientific
Varování

PC520792-10G PC520792-1G PC520792-5G
APOSPC520792-10GEA 29830 CZK
APOSPC520792-10G APOSPC520792-1G APOSPC520792-5G
2,3-Dimethylbenzotrifluoride 97%
2,3-Dimethylbenzotrifluoride
Vzorec: C₉H₉F₃
Storage Temperature: Ambient
MDL Number: MFCD11226579
CAS číslo: 80245-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR