Tisk…

3-(Trifluoromethoxy)pyrrolidine hydrochloride 97%

Dodavatel: Apollo Scientific

PC520779-1G PC520779-250MG
APOSPC520779-1GEA 19540 CZK
APOSPC520779-1G APOSPC520779-250MG
3-(Trifluoromethoxy)pyrrolidine hydrochloride 97%
3-(Trifluoromethoxy)pyrrolidine hydrochloride
Vzorec: C₅H₉ClF₃NO
Bod tání: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD23381937
CAS číslo: 1246466-85-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR