Tisk…

2-Chloro-3-(difluoromethyl)-6-methylpyridine 95%

Dodavatel: Apollo Scientific

PC520756-1G PC520756-500MG
APOSPC520756-1GEA 10580 CZK
APOSPC520756-1G APOSPC520756-500MG
2-Chloro-3-(difluoromethyl)-6-methylpyridine 95%
2-Chloro-3-(difluoromethyl)-6-methylpyridine
Vzorec: C₇H₆ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD22199315
CAS číslo: 1374659-36-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR