Tisk…

A-967079 95%

Dodavatel: Apollo Scientific

Synonyms: (1E,3E)-1-(4-Fluorophenyl)-2-methyl-1-penten-3-one oxime, A 967079

PC520753-100MG PC520753-250MG PC520753-500MG
APOSPC520753-100MGEA 9290 CZK
APOSPC520753-100MG APOSPC520753-250MG APOSPC520753-500MG
A-967079 95%
A-967079
Vzorec: C₁₂H₁₄FNO
MW: 207,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20488058
CAS číslo: 1170613-55-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR