Tisk…

6-Chloro-8-fluoroquinoline 95%

Dodavatel: Apollo Scientific

PC520731-10G PC520731-1G PC520731-5G
APOSPC520731-10GEA 32800 CZK
APOSPC520731-10G APOSPC520731-1G APOSPC520731-5G
6-Chloro-8-fluoroquinoline 95%
6-Chloro-8-fluoroquinoline
Vzorec: C₉H₅ClFN
MW: 181.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658595
CAS číslo: 52200-53-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR