1-(Difluorophenylmethyl)-4-methylbenzene 97%
Dodavatel: Apollo Scientific
PC520726-1G
PC520726-5G
PC520726-25G
APOSPC520726-1GEA
2480
CZK
APOSPC520726-1G
APOSPC520726-5G
APOSPC520726-25G
1-(Difluorophenylmethyl)-4-methylbenzene 97%
1-(Difluorophenylmethyl)-4-methylbenzene
Vzorec:
C₁₄H₁₂F₂ Storage Temperature: Ambient |
MDL Number:
MFCD16140157 CAS číslo: 174075-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...