Tisk…

2-(Difluoromethyl)-4-fluoro-1-nitrobenzene 95%

Dodavatel: Apollo Scientific

PC520712-10G PC520712-1G PC520712-5G
APOSPC520712-10GEA 29490 CZK
APOSPC520712-10G APOSPC520712-1G APOSPC520712-5G
2-(Difluoromethyl)-4-fluoro-1-nitrobenzene 95%
2-(Difluoromethyl)-4-fluoro-1-nitrobenzene
Vzorec: C₇H₄F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD14698664
CAS číslo: 1214333-17-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR