Tisk…

1-(Difluoromethyl)-3,5-bis(trifluoromethyl)benzene 97%

Dodavatel: Apollo Scientific

PC520710-10G PC520710-1G PC520710-5G
APOSPC520710-10GEA 27510 CZK
APOSPC520710-10G APOSPC520710-1G APOSPC520710-5G
1-(Difluoromethyl)-3,5-bis(trifluoromethyl)benzene 97%
1-(Difluoromethyl)-3,5-bis(trifluoromethyl)benzene
Vzorec: C₉H₄F₈
MW: 264.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698530
CAS číslo: 1214388-66-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR