Tisk…

1-Nitro-4-(2,2,2-trifluoro-1-methylethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520703-10G PC520703-1G PC520703-5G
APOSPC520703-10GEA 32800 CZK
APOSPC520703-10G APOSPC520703-1G APOSPC520703-5G
1-Nitro-4-(2,2,2-trifluoro-1-methylethyl)benzene 95%
1-Nitro-4-(2,2,2-trifluoro-1-methylethyl)benzene
Vzorec: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD14525520
CAS číslo: 1221272-94-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR