Tisk…

1-Nitro-2-(2,2,2-trifluoroethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520702-10G PC520702-1G PC520702-5G
APOSPC520702-10GEA 17000 CZK
APOSPC520702-10G APOSPC520702-1G APOSPC520702-5G
1-Nitro-2-(2,2,2-trifluoroethyl)benzene 95%
1-Nitro-2-(2,2,2-trifluoroethyl)benzene
Vzorec: C₈H₆F₃NO₂
MW: 205.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14525517
CAS číslo: 145914-06-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR