Tisk…

1-(Difluoromethoxy)-3-fluorobenzene 95%

Dodavatel: Apollo Scientific

PC520691-10G PC520691-1G PC520691-5G
APOSPC520691-10GEA 46290 CZK
APOSPC520691-10G APOSPC520691-1G APOSPC520691-5G
1-(Difluoromethoxy)-3-fluorobenzene 95%
1-(Difluoromethoxy)-3-fluorobenzene
Vzorec: C₇H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD13194315
CAS číslo: 34888-08-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR