2-Methyl-3-(trifluoromethyl)pyridine 97%
Dodavatel: Apollo Scientific
PC520681-10G
PC520681-1G
PC520681-5G
APOSPC520681-10GEA
26520
CZK
APOSPC520681-10G
APOSPC520681-1G
APOSPC520681-5G
2-Methyl-3-(trifluoromethyl)pyridine 97%
2-Methyl-3-(trifluoromethyl)pyridine
Vzorec:
C₇H₆F₃N Storage Temperature: Ambient |
MDL Number:
MFCD12546975 CAS číslo: 106877-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...