Tisk…

4-Chloro-1,2-bis-(trifluoromethyl)benzene 97%

Dodavatel: Apollo Scientific

PC520673-10G PC520673-1G PC520673-5G
APOSPC520673-10GEA 28500 CZK
APOSPC520673-10G APOSPC520673-1G APOSPC520673-5G
4-Chloro-1,2-bis-(trifluoromethyl)benzene 97%
4-Chloro-1,2-bis-(trifluoromethyl)benzene
Vzorec: C₈H₃ClF₆
Storage Temperature: Ambient
MDL Number: MFCD11100533
CAS číslo: 320-30-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR