1-Fluoro-3-(2,2,2-trifluoroethyl)benzene 97%
Dodavatel: Apollo Scientific
PC520671-10G
PC520671-5G
APOSPC520671-10GEA
19870
CZK
APOSPC520671-10G
APOSPC520671-5G
1-Fluoro-3-(2,2,2-trifluoroethyl)benzene 97%
1-Fluoro-3-(2,2,2-trifluoroethyl)benzene
Vzorec:
C₈H₆F₄ Storage Temperature: Ambient |
MDL Number:
MFCD11100171 CAS číslo: 81577-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...