Tisk…

3-Chloro-2,6-difluorobenzotrifluoride 95%

Dodavatel: Apollo Scientific

PC520647-10G PC520647-1G PC520647-5G
APOSPC520647-10GEA 25860 CZK
APOSPC520647-10G APOSPC520647-1G APOSPC520647-5G
3-Chloro-2,6-difluorobenzotrifluoride 95%
3-Chloro-2,6-difluorobenzotrifluoride
Vzorec: C₇H₂ClF₅
Storage Temperature: Ambient
MDL Number: MFCD11226546
CAS číslo: 1099597-36-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR