Tisk…

1-Bromo-3-(3,3,3-trifluoropropyl)benzene 95%

Dodavatel: Apollo Scientific

PC520644-10G PC520644-1G PC520644-5G
APOSPC520644-10GEA 35050 CZK
APOSPC520644-10G APOSPC520644-1G APOSPC520644-5G
1-Bromo-3-(3,3,3-trifluoropropyl)benzene 95%
1-Bromo-3-(3,3,3-trifluoropropyl)benzene
Vzorec: C₉H₈BrF₃
Storage Temperature: Ambient
MDL Number: MFCD11226672
CAS číslo: 1099597-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR