1-Bromo-3-(2,2-difluoropropyl)benzene 97%
Dodavatel: Apollo Scientific
PC520641-1G
PC520641-5G
APOSPC520641-1GEA
7310
CZK
APOSPC520641-1G
APOSPC520641-5G
1-Bromo-3-(2,2-difluoropropyl)benzene 97%
1-Bromo-3-(2,2-difluoropropyl)benzene
Vzorec:
C₉H₉BrF₂ Storage Temperature: Ambient |
MDL Number:
MFCD11226516 CAS číslo: 1099598-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...