Tisk…

Methyl-5-fluoropicolinate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 5-fluoropicolinate, Methyl 5-fluoropyridine-2-carboxylate

PC520639-1G PC520639-5G
APOSPC520639-1GEA 1080 CZK
APOSPC520639-1G APOSPC520639-5G
Methyl-5-fluoropicolinate 98%
Methyl-5-fluoropicolinate
Vzorec: C₇H₆FNO₂
MW: 155,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845630
CAS číslo: 107504-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR