Tisk…

1-(2,2,2-Trifluoroethyl)-3-(trifluoromethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520629-10G PC520629-1G PC520629-5G
APOSPC520629-10GEA 26190 CZK
APOSPC520629-10G APOSPC520629-1G APOSPC520629-5G
1-(2,2,2-Trifluoroethyl)-3-(trifluoromethyl)benzene 95%
1-(2,2,2-Trifluoroethyl)-3-(trifluoromethyl)benzene
Vzorec: C₉H₆F₆
Storage Temperature: Ambient
MDL Number: MFCD11226548
CAS číslo: 50562-22-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR