Tisk…

6-Chloro-2-fluoro-3-methylbenzotrifluoride 97%

Dodavatel: Apollo Scientific

PC520623-1G PC520623-5G
APOSPC520623-1GEA 5860 CZK
APOSPC520623-1G APOSPC520623-5G
6-Chloro-2-fluoro-3-methylbenzotrifluoride 97%
6-Chloro-2-fluoro-3-methylbenzotrifluoride
Vzorec: C₈H₅ClF₄
MW: 212.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226667
CAS číslo: 1099597-48-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR