2-Chloro-6-methyl-3-(trifluoromethyl)pyridine 97%
Dodavatel: Apollo Scientific
PC520613-10G
PC520613-1G
PC520613-5G
APOSPC520613-10GEA
32800
CZK
APOSPC520613-10G
APOSPC520613-1G
APOSPC520613-5G
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine 97%
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine
Vzorec:
C₇H₅ClF₃N Storage Temperature: Ambient |
MDL Number:
MFCD11226678 CAS číslo: 1099597-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...