1-Bromo-4-(2,2-difluoropropyl)benzene 97%
Dodavatel: Apollo Scientific
PC520609-10G
PC520609-1G
PC520609-5G
APOSPC520609-10GEA
42980
CZK
APOSPC520609-10G
APOSPC520609-1G
APOSPC520609-5G
1-Bromo-4-(2,2-difluoropropyl)benzene 97%
1-Bromo-4-(2,2-difluoropropyl)benzene
Vzorec:
C₉H₉BrF₂ Storage Temperature: Ambient |
MDL Number:
MFCD11226517 CAS číslo: 1099598-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...