Tisk…

2,4-Bis(trifluoromethyl)pyrimidine-5-carboxylic acid 97%

Dodavatel: Apollo Scientific

PC520603-1G PC520603-500MG PC520603-5G
APOSPC520603-1GEA 7310 CZK
APOSPC520603-1G APOSPC520603-500MG APOSPC520603-5G
2,4-Bis(trifluoromethyl)pyrimidine-5-carboxylic acid 97%
2,4-Bis(trifluoromethyl)pyrimidine-5-carboxylic acid
Vzorec: C₇H₂F₆N₂O₂
MW: 260,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697912
CAS číslo: 188781-46-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR