Tisk…

3,6-Difluoro-2-(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC520593-1G PC520593-5G
APOSPC520593-1GEA 4570 CZK
APOSPC520593-1G APOSPC520593-5G
3,6-Difluoro-2-(trifluoromethyl)pyridine 95%
3,6-Difluoro-2-(trifluoromethyl)pyridine
Vzorec: C₆H₂F₅N
Bod varu: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD10699122
CAS číslo: 1099597-92-8
UN: 1992
ADR: 3+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR