Tisk…

4-Bromo-2-(trifluoromethoxy)anisole 95%

Dodavatel: Apollo Scientific

PC520587-10G PC520587-5G
APOSPC520587-10GEA 15250 CZK
APOSPC520587-10G APOSPC520587-5G
4-Bromo-2-(trifluoromethoxy)anisole 95%
4-Bromo-2-(trifluoromethoxy)anisole
Vzorec: C₈H₆BrF₃O₂
MW: 271.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907950
CAS číslo: 853771-88-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR