Tisk…

7-Fluoro-2,3,4,9-tetrahydro-1H-β-carboline 95%

Dodavatel: Apollo Scientific

PC520580-1G PC520580-250MG PC520580-500MG PC520580-100MG
APOSPC520580-1GEA 26590 CZK
APOSPC520580-1G APOSPC520580-250MG APOSPC520580-500MG APOSPC520580-100MG
7-Fluoro-2,3,4,9-tetrahydro-1H-β-carboline 95%
7-Fluoro-2,3,4,9-tetrahydro-1H-beta-carboline
Vzorec: C₁₁H₁₁FN₂
Storage Temperature: Ambient
MDL Number: MFCD09264527
CAS číslo: 177858-80-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR