4-(Difluoromethyl)-1-fluorobenzene 97%
Dodavatel: Apollo Scientific
PC520555-25G
PC520555-5G
APOSPC520555-25GEA
16570
CZK
APOSPC520555-25G
APOSPC520555-5G
4-(Difluoromethyl)-1-fluorobenzene 97%
4-(Difluoromethyl)-1-fluorobenzene
Vzorec:
C₇H₅F₃ Storage Temperature: Ambient |
MDL Number:
MFCD07368983 CAS číslo: 26132-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...