Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate 97%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
PC520551-1G
PC520551-5G
APOSPC520551-1GEA
4830
CZK
APOSPC520551-1G
APOSPC520551-5G
Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate 97%
Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate
Vzorec:
C₉H₉F₃N₂O₂ MW: 234,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07367386 CAS číslo: 306960-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...