3-Amino-4-fluorophenylboronic acid pinacol ester 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Amino-4-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Amino-4-fluorophenylboronic acid, pinacol ester
PC52054-1G
PC52054-5G
APOSPC52054-1GEA
1620
CZK
APOSPC52054-1G
APOSPC52054-5G
3-Amino-4-fluorophenylboronic acid pinacol ester 96%
3-Amino-4-fluorophenylboronic acid pinacol ester
Vzorec:
C₁₂H₁₇BFNO₂ MW: 237,08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11044430 CAS číslo: 1003575-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...