Tisk…

2,5-Diamino-3-fluoro-4-(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific

PC520529-1G PC520529-250MG
APOSPC520529-1GEA 10620 CZK
APOSPC520529-1G APOSPC520529-250MG
2,5-Diamino-3-fluoro-4-(trifluoromethyl)pyridine 95%
2,5-Diamino-3-fluoro-4-(trifluoromethyl)pyridine
Vzorec: C₆H₅F₄N₃
Storage Temperature: Ambient
MDL Number: MFCD04038327
CAS číslo: 675602-90-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR