Tisk…

(5-Fluoro-3-pyridyl)methanol 97%

Dodavatel: Apollo Scientific

Synonyms: (5-Fluoropyridin-3-yl)methanol

PC520517-10G PC520517-1G PC520517-5G
APOSPC520517-10GEA 28170 CZK
APOSPC520517-10G APOSPC520517-1G APOSPC520517-5G
(5-Fluoro-3-pyridyl)methanol 97%
(5-Fluoro-3-pyridyl)methanol
Vzorec: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD01758680
CAS číslo: 22620-32-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR