1-Bromo-4-[(trifluoromethyl)sulphonyl]benzene 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-4-((trifluoromethyl)sulphonyl)benzene
PC520507-10G
PC520507-1G
PC520507-5G
APOSPC520507-10GEA
34390
CZK
APOSPC520507-10G
APOSPC520507-1G
APOSPC520507-5G
1-Bromo-4-[(trifluoromethyl)sulphonyl]benzene 96%
1-Bromo-4-[(trifluoromethyl)sulphonyl]benzene
Vzorec:
C₇H₄BrF₃O₂S MW: 289,08 g/mol Bod tání: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00474580 CAS číslo: 312-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...